siRNA Details
Gene Symbol: Pnkp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004259.1 | NP_001004259.1 | 6 | 782 | ||
XM_006229058.2 | XP_006229120.1 | 4 | 1076 | |||
XM_008759371.1 | XP_008757593.1 | 5 | 1164 | |||
GenBank mRNA | BC079307.1 | - | 6 | 782 | 7134659 |
