siRNA Details
Gene Symbol: CDC42BPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003607.3 | NP_003598.2 | 2 | 1194 | ||
NM_014826.4 | NP_055641.3 | 2 | 1194 | |||
XM_005273317.3 | XP_005273374.1 | 2 | 1306 | |||
XM_005273318.3 | XP_005273375.1 | 2 | 1306 | |||
XM_005273320.3 | XP_005273377.1 | 2 | 1402 | |||
XM_005273321.3 | XP_005273378.1 | 2 | 1306 | |||
XM_005273322.3 | XP_005273379.1 | 2 | 1306 | |||
XM_005273324.3 | XP_005273381.1 | 2 | 1402 | |||
XM_006711834.3 | XP_006711897.1 | 2 | 1306 | |||
XM_006711835.3 | XP_006711898.1 | 2 | 1306 | |||
XM_011544306.2 | XP_011542608.1 | 2 | 1561 | |||
XM_011544307.2 | XP_011542609.1 | 2 | 1306 | |||
XM_011544308.2 | XP_011542610.1 | 2 | 1306 | |||
XM_011544309.2 | XP_011542611.1 | 2 | 1306 | |||
XM_017002578.1 | XP_016858067.1 | 2 | 1306 | |||
XM_017002579.1 | XP_016858068.1 | 2 | 1561 | |||
XM_017002581.1 | XP_016858070.1 | 2 | 1561 | |||
XM_017002582.1 | XP_016858071.1 | 2 | 1306 | |||
GenBank mRNA | AB208968.1 | - | 2 | 909 | ||
AJ518975.1 | - | 2 | 251 | |||
AJ518976.1 | - | 2 | 251 | |||
AY166586.1 | - | 2 | 251 | |||
BC136333.1 | - | 2 | 278 | 9020320 | ||
BQ232716.1 | - | 2 | 357 | |||
U59305.1 | - | 2 | 1539 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
