siRNA Details
Gene Symbol: Mmadhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004280.1 | NP_001004280.1 | 4 | 396 | ||
XM_006234158.2 | XP_006234220.1 | 4 | 515 | |||
GenBank mRNA | BC078953.1 | - | 4 | 396 | 7110154 | |
FQ212704.1 | - | 4 | 391 | |||
FQ216239.1 | - | 4 | 391 | |||
FQ219117.1 | - | 4 | 392 | |||
FQ228704.1 | - | 4 | 392 | |||
FQ229971.1 | - | 4 | 361 |
