siRNA Details
Gene Symbol: FLJ42258 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124252.1 | - | 1 | 983 | ||
BC133014.1 | - | 1 | 638 | |||
BC133018.1 | - | 1 | 638 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FLJ42258 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124252.1 | - | 1 | 983 | ||
BC133014.1 | - | 1 | 638 | |||
BC133018.1 | - | 1 | 638 |