siRNA Details
Gene Symbol: TRIM67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004342.3 | NP_001004342.3 | 10 | 7392 | ||
NM_001300889.1 | NP_001287818.1 | 12 | 7206 | |||
XM_011544192.2 | XP_011542494.1 | 10 | 7386 | |||
GenBank mRNA | AK055338.1 | - | 1 | 2430 | ||
AK126782.1 | - | 11 | 2799 |
