siRNA Details
Gene Symbol: FLJ16171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046113.1 | 1 | 185 | |||
GenBank mRNA | AK131247.1 | - | 1 | 185 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FLJ16171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046113.1 | 1 | 185 | |||
GenBank mRNA | AK131247.1 | - | 1 | 185 |