siRNA Details
Gene Symbol: 2610008E11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004362.2 | NP_001004362.2 | 6 | 3126 | ||
GenBank mRNA | AK011343.1 | - | 1 | 951 | ||
AK082019.1 | - | 1 | 468 | |||
BC082302.1 | - | 6 | 3083 | 6838422 |
