siRNA Details
Gene Symbol: OR6C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005182.1 | NP_001005182.1 | 1 | 226 | ||
GenBank mRNA | BC136957.1 | - | 1 | 265 | 9020949 | |
BC136965.1 | - | 1 | 264 | 9020957 |
