siRNA Details
Gene Symbol: Zfp960 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005358.2 | NP_001005358.2 | 4 | 2556 | ||
NM_001163919.1 | NP_001157391.1 | 5 | 1379 | |||
GenBank mRNA | AK145357.1 | - | 5 | 1367 | ||
BC018101.1 | - | 4 | 2546 | 3494875 | ||
BC064743.1 | - | 5 | 875 | |||
BC066177.1 | - | 5 | 875 |
Gene Symbol: Zfp97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011765.5 | NP_035895.3 | 4 | 2607 | ||
GenBank mRNA | DV660800.1 | - | 1 | 438 | ||
L20450.1 | - | 2659 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
