siRNA Details
Gene Symbol: Ado | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005419.2 | NP_001005419.2 | 1 | 3130 | ||
GenBank mRNA | AK047542.1 | - | 1 | 1691 | ||
BC057106.1 | - | 1 | 3116 | 5721417 | ||
BC058407.1 | - | 1 | 3116 | 5721417 |
