siRNA Details
Gene Symbol: Fam160a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005538.2 | NP_001005538.2 | 12 | 3123 | ||
XM_006229931.2 | XP_006229993.1 | 12 | 3115 | |||
XM_006229932.2 | XP_006229994.1 | 12 | 2562 | |||
GenBank mRNA | BC082011.1 | - | 12 | 2515 | 7132039 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001242364, NM_001242365, NM_199009, NM_001242363, BC021532, and others.
Gene Ontology Categories:
