siRNA Details
Gene Symbol: Mavs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005556.1 | NP_001005556.1 | 6 | 1183 | ||
XM_006235034.2 | XP_006235096.1 | 6 | 1051 | |||
XM_006235035.2 | XP_006235097.1 | 6 | 1765 | |||
XM_006235036.2 | XP_006235098.1 | 7 | 1084 | |||
XM_006235037.2 | XP_006235099.1 | 5 | 1087 | |||
XM_008762179.1 | XP_008760401.1 | 6 | 2258 | |||
XM_008762180.1 | XP_008760402.1 | 6 | 1071 | |||
GenBank mRNA | BC081869.1 | - | 6 | 1183 | 7106026 |
