siRNA Details
Gene Symbol: Akap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005557.1 | NP_001005557.1 | 2, 3 | 180 | ||
XM_006237488.2 | XP_006237550.1 | 3, 4 | 365 | |||
XM_006237489.2 | XP_006237551.1 | 4, 5 | 476 | |||
XM_006237491.2 | XP_006237553.1 | 3, 4 | 622 | |||
XM_006237492.2 | XP_006237554.1 | 3, 4 | 249 | |||
XM_008763287.1 | XP_008761509.1 | 3, 4 | 212 | |||
XM_008763288.1 | XP_008761510.1 | 3, 4 | 284 | |||
XM_008763289.1 | XP_008761511.1 | 2, 3 | 183 | |||
XM_008763290.1 | XP_008761512.1 | 2, 3 | 162 | |||
GenBank mRNA | BC081924.1 | - | 2, 3 | 180 | 7112940 |
