siRNA Details
Gene Symbol: Akap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005557.1 | NP_001005557.1 | 3 | 385 | ||
XM_006237488.2 | XP_006237550.1 | 4 | 570 | |||
XM_006237489.2 | XP_006237551.1 | 5 | 681 | |||
XM_006237491.2 | XP_006237553.1 | 4 | 827 | |||
XM_006237492.2 | XP_006237554.1 | 4 | 454 | |||
XM_008763287.1 | XP_008761509.1 | 4 | 417 | |||
XM_008763288.1 | XP_008761510.1 | 4 | 489 | |||
XM_008763289.1 | XP_008761511.1 | 3 | 388 | |||
XM_008763290.1 | XP_008761512.1 | 3 | 367 | |||
GenBank mRNA | BC081924.1 | - | 3 | 385 | 7112940 |
