siRNA Details
Gene Symbol: FAM21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005751.2 | NP_001005751.1 | 8, 9 | 855 | ||
NM_001291398.1 | NP_001278327.1 | 8, 9 | 855 | |||
XM_005269806.3 | XP_005269863.2 | 8, 9 | 803 | |||
XM_005269807.3 | XP_005269864.2 | 8, 9 | 803 | |||
XM_005269808.3 | XP_005269865.2 | 8, 9 | 803 | |||
XM_006717831.3 | XP_006717894.2 | 8, 9 | 803 | |||
XM_006717832.3 | XP_006717895.2 | 8, 9 | 803 | |||
XM_011539786.2 | XP_011538088.1 | 8, 9 | 803 | |||
XM_017016227.1 | XP_016871716.1 | 8, 9 | 803 | |||
XM_017016228.1 | XP_016871717.1 | 8, 9 | 803 | |||
XM_017016229.1 | XP_016871718.1 | 8, 9 | 803 | |||
XM_017016230.1 | XP_016871719.1 | 7, 8 | 668 | |||
XM_017016231.1 | XP_016871720.1 | 8, 9 | 803 | |||
XM_017016232.1 | XP_016871721.1 | 8, 9 | 803 | |||
XM_017016233.1 | XP_016871722.1 | 5, 6 | 378 | |||
XM_017016234.1 | XP_016871723.1 | 9, 10 | 897 | |||
GenBank mRNA | AK001686.1 | - | 5, 6 | 506 | ||
AK295129.1 | - | 8, 9 | 807 | |||
AK304116.1 | - | 8, 9 | 791 | |||
AK311628.1 | - | 5, 6 | 409 | |||
BC075815.1 | - | 5, 6 | 420 | |||
BC082258.1 | - | 8, 9 | 782 | 6278433 | ||
BC144479.1 | - | 8, 9 | 884 | 9053006 | ||
DA402580.1 | - | 13 | ||||
DC403380.1 | - | 5, 6 | 409 |
Gene Symbol: FAM21C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC150611.1 | - | 8, 9 | 820 | 9056981 | |
BM126842.1 | - | 4, 5 | 310 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
