siRNA Details
Gene Symbol: FAM21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005751.2 | NP_001005751.1 | 22 | 2376 | ||
NM_001291398.1 | NP_001278327.1 | 22 | 2376 | |||
XM_005269806.3 | XP_005269863.2 | 21 | 2252 | |||
XM_005269807.3 | XP_005269864.2 | 21 | 2237 | |||
XM_005269808.3 | XP_005269865.2 | 22 | 2324 | |||
XM_005269810.3 | XP_005269867.2 | 4 | 469 | |||
XM_011539786.2 | XP_011538088.1 | 22 | 2324 | |||
XM_017016227.1 | XP_016871716.1 | 22 | 2324 | |||
XM_017016228.1 | XP_016871717.1 | 21 | 2252 | |||
XM_017016229.1 | XP_016871718.1 | 21 | 2237 | |||
XM_017016230.1 | XP_016871719.1 | 21 | 2189 | |||
XM_017016232.1 | XP_016871721.1 | 20 | 2165 | |||
XM_017016233.1 | XP_016871722.1 | 19 | 1899 | |||
XM_017016234.1 | XP_016871723.1 | 21 | 2259 | |||
XM_017016235.1 | XP_016871724.1 | 13 | 1275 | |||
GenBank mRNA | AK001686.1 | - | 19 | 2028 | ||
AK304116.1 | - | 22 | 2312 | |||
BC075815.1 | - | 19 | 1941 | |||
BC082258.1 | - | 22 | 2303 | 6278433 | ||
BC144479.1 | - | 22 | 2405 | 9053006 | ||
CX762870.1 | - | 1 | 132 | |||
DA466642.1 | - | 4 | 527 |
Gene Symbol: FAM21C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169106.1 | NP_001162577.1 | 22 | 2376 | ||
NM_015262.2 | NP_056077.2 | 22 | 2376 | |||
XM_011539566.1 | XP_011537868.1 | 22 | 2343 | |||
XM_011539567.1 | XP_011537869.1 | 21 | 2271 | |||
XM_011539568.1 | XP_011537870.1 | 21 | 2256 | |||
XM_011539570.2 | XP_011537872.1 | 21 | 2232 | |||
XM_011539571.1 | XP_011537873.1 | 22 | 2343 | |||
XM_011539573.2 | XP_011537875.1 | 4 | 469 | |||
XM_017016014.1 | XP_016871503.1 | 22 | 2343 | |||
XM_017016015.1 | XP_016871504.1 | 21 | 2271 | |||
XM_017016016.1 | XP_016871505.1 | 21 | 2256 | |||
XM_017016017.1 | XP_016871506.1 | 20 | 2184 | |||
XM_017016020.1 | XP_016871509.1 | 21 | 2271 | |||
XM_017016021.1 | XP_016871510.1 | 22 | 2271 | |||
XM_017016022.1 | XP_016871511.1 | 22 | 2317 | |||
XM_017016023.1 | XP_016871512.1 | 21 | 2189 | |||
XM_017016024.1 | XP_016871513.1 | 10 | 1085 | |||
XM_017016025.1 | XP_016871514.1 | 11 | 1156 | |||
GenBank mRNA | AB011164.1 | - | 22 | 2357 | ||
AK001545.1 | - | 4 | 537 | |||
AK291430.1 | - | 22 | 2364 | |||
AK293929.1 | - | 17 | 1827 | |||
BC150611.1 | - | 22 | 2341 | 9056981 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
