siRNA Details
Gene Symbol: FAM21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005751.2 | NP_001005751.1 | 31 | 4184 | ||
NM_001291398.1 | NP_001278327.1 | 30 | 4121 | |||
XM_005269806.3 | XP_005269863.2 | 30 | 4060 | |||
XM_005269807.3 | XP_005269864.2 | 30 | 4045 | |||
XM_005269808.3 | XP_005269865.2 | 29 | 3946 | |||
XM_005269810.3 | XP_005269867.2 | 13 | 2277 | |||
XM_006717831.3 | XP_006717894.2 | 30 | 3979 | |||
XM_006717832.3 | XP_006717895.2 | 28 | 3844 | |||
XM_011539786.2 | XP_011538088.1 | 28 | 3733 | |||
XM_017016227.1 | XP_016871716.1 | 30 | 4009 | |||
XM_017016228.1 | XP_016871717.1 | 29 | 3997 | |||
XM_017016229.1 | XP_016871718.1 | 29 | 3982 | |||
XM_017016230.1 | XP_016871719.1 | 30 | 3997 | |||
XM_017016231.1 | XP_016871720.1 | 29 | 3916 | |||
XM_017016232.1 | XP_016871721.1 | 28 | 3910 | |||
XM_017016233.1 | XP_016871722.1 | 28 | 3707 | |||
XM_017016234.1 | XP_016871723.1 | 30 | 4067 | |||
XM_017016235.1 | XP_016871724.1 | 22 | 3083 | |||
GenBank mRNA | AK055424.1 | - | 1 | 1743 | ||
AK125147.1 | - | 3 | 2538 | |||
AK304116.1 | - | 31 | 4120 | |||
AL080183.1 | - | 7 | 1516 | |||
BC065500.1 | - | 3 | 422 | |||
BC075815.1 | - | 27 | 3686 | |||
BC082258.1 | - | 31 | 4111 | 6278433 | ||
BC144479.1 | - | 30 | 4150 | 9053006 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
