siRNA Details
Gene Symbol: ZNF780B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005851.2 | NP_001005851.1 | 4 | 778 | ||
XM_005258590.4 | XP_005258647.1 | 6 | 1010 | |||
XM_005258591.4 | XP_005258648.1 | 5 | 3599 | |||
XM_005258592.3 | XP_005258649.1 | 6 | 918 | |||
XM_005258593.2 | XP_005258650.1 | 6 | 994 | |||
XM_005258595.3 | XP_005258652.1 | 5 | 844 | |||
XM_006723072.3 | XP_006723135.1 | 6 | 3760 | |||
XM_011526593.2 | XP_011524895.1 | 5 | 790 | |||
XM_017026425.1 | XP_016881914.1 | 6 | 3757 | |||
XM_017026426.1 | XP_016881915.1 | 6 | 862 | |||
XM_017026427.1 | XP_016881916.1 | 5 | 841 | |||
GenBank mRNA | AK127063.1 | - | 6 | 971 | ||
BC136960.1 | - | 4 | 758 | 9020952 |
Gene Symbol: ZNF780A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010880.2 | NP_001010880.2 | 6 | 1358 | ||
NM_001142577.1 | NP_001136049.1 | 6 | 1284 | |||
NM_001142578.1 | NP_001136050.1 | 6 | 1281 | |||
XM_005258773.2 | XP_005258830.1 | 6 | 1361 | |||
XM_006723150.3 | XP_006723213.1 | 5 | 1981 | |||
XM_011526771.2 | XP_011525073.1 | 6 | 1403 | |||
XM_011526772.2 | XP_011525074.1 | 6 | 1415 | |||
XM_017026611.1 | XP_016882100.1 | 6 | 2068 | |||
XM_017026612.1 | XP_016882101.1 | 7 | 2207 | |||
XM_017026613.1 | XP_016882102.1 | 7 | 1570 | |||
XM_017026614.1 | XP_016882103.1 | 7 | 1558 | |||
XM_017026615.1 | XP_016882104.1 | 7 | 1512 | |||
XM_017026616.1 | XP_016882105.1 | 7 | 1436 | |||
XM_017026617.1 | XP_016882106.1 | 5 | 1910 | |||
XM_017026618.1 | XP_016882107.1 | 6 | 1408 | |||
XM_017026619.1 | XP_016882108.1 | 5 | 1974 | |||
GenBank mRNA | AK091274.1 | - | 6 | 1343 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
