siRNA Details
Gene Symbol: ZNF780B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005851.2 | NP_001005851.1 | 4 | 326 | ||
XM_005258590.4 | XP_005258647.1 | 6 | 558 | |||
XM_005258591.4 | XP_005258648.1 | 5 | 3147 | |||
XM_005258592.3 | XP_005258649.1 | 6 | 466 | |||
XM_005258593.2 | XP_005258650.1 | 6 | 542 | |||
XM_005258595.3 | XP_005258652.1 | 5 | 392 | |||
XM_006723072.3 | XP_006723135.1 | 6 | 3308 | |||
XM_011526593.2 | XP_011524895.1 | 5 | 338 | |||
XM_017026425.1 | XP_016881914.1 | 6 | 3305 | |||
XM_017026426.1 | XP_016881915.1 | 6 | 410 | |||
XM_017026427.1 | XP_016881916.1 | 5 | 389 | |||
GenBank mRNA | AK127063.1 | - | 6 | 519 | ||
BC136960.1 | - | 4 | 306 | 9020952 | ||
CB053507.1 | - | 4 | 334 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
