siRNA Details
Gene Symbol: CEP162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286206.1 | NP_001273135.1 | 5 | 479 | ||
NM_014895.3 | NP_055710.2 | 5 | 533 | |||
XM_005248674.3 | XP_005248731.1 | 4 | 656 | |||
XM_005248678.3 | XP_005248735.1 | 5 | 787 | |||
XM_006715380.2 | XP_006715443.1 | 6 | 627 | |||
XM_011535594.2 | XP_011533896.1 | 5 | 787 | |||
XM_017010483.1 | XP_016865972.1 | 4 | 1212 | |||
XM_017010484.1 | XP_016865973.1 | 6 | 830 | |||
GenBank mRNA | AB023226.1 | - | 5 | 440 | ||
AK308190.1 | - | 5 | 500 | |||
BC063632.1 | - | 5 | 506 | |||
DC424230.1 | - | 5 | 515 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_199316, BC151006, AK163175, AK135482, BC004073, and others.
Gene Ontology Categories:
