siRNA Details
Gene Symbol: DLGAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014902.5 | NP_055717.2 | 3 | 1476 | ||
XM_011528688.2 | XP_011526990.1 | 2 | 1242 | |||
XM_011528689.2 | XP_011526991.1 | 2 | 1249 | |||
GenBank mRNA | AB023181.1 | - | 3 | 1418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
