siRNA Details
Gene Symbol: ZNF473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017026574.1 | XP_016882063.1 | 3 | 1847 | ||
GenBank mRNA | AB032967.1 | - | 5 | 604 | ||
AF454744.1 | - | 4 | 559 | |||
AK292452.1 | - | 5 | 497 | |||
AK302293.1 | - | 4 | 454 | |||
BC018612.1 | - | 5 | 527 | 4634056 | ||
BX648093.1 | - | 5 | 648 |
