siRNA Details
Gene Symbol: KIAA0556 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015202.3 | NP_056017.3 | 23 | 4307 | ||
XM_005255201.3 | XP_005255258.1 | 24 | 4509 | |||
XM_005255202.3 | XP_005255259.1 | 24 | 4455 | |||
XM_005255203.3 | XP_005255260.1 | 23 | 4380 | |||
XM_005255208.2 | XP_005255265.1 | 14 | 3207 | |||
XM_006721025.3 | XP_006721088.1 | 22 | 4248 | |||
XM_011545773.2 | XP_011544075.1 | 24 | 4479 | |||
XM_011545774.1 | XP_011544076.1 | 23 | 4425 | |||
XM_011545775.2 | XP_011544077.1 | 23 | 4389 | |||
XM_011545776.2 | XP_011544078.1 | 23 | 4377 | |||
XM_011545777.2 | XP_011544079.1 | 10 | 2640 | |||
XM_017023085.1 | XP_016878574.1 | 22 | 4260 | |||
XM_017023086.1 | XP_016878575.1 | 22 | 4260 | |||
XM_017023087.1 | XP_016878576.1 | 22 | 4194 | |||
XM_017023088.1 | XP_016878577.1 | 19 | 3981 | |||
GenBank mRNA | AB011128.2 | - | 23 | 4305 | ||
BC150276.1 | - | 23 | 4305 | 9007250 |
