siRNA Details
Gene Symbol: KIAA0556 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015202.3 | NP_056017.3 | 27, 28 | 4833 | ||
XM_005255201.3 | XP_005255258.1 | 28, 29 | 5035 | |||
XM_005255202.3 | XP_005255259.1 | 28, 29 | 4981 | |||
XM_005255203.3 | XP_005255260.1 | 27, 28 | 4906 | |||
XM_005255208.2 | XP_005255265.1 | 18, 19 | 3733 | |||
XM_006721025.3 | XP_006721088.1 | 26, 27 | 4774 | |||
XM_011545773.2 | XP_011544075.1 | 28, 29 | 5005 | |||
XM_011545774.1 | XP_011544076.1 | 27, 28 | 4951 | |||
XM_011545775.2 | XP_011544077.1 | 27, 28 | 4915 | |||
XM_011545776.2 | XP_011544078.1 | 27, 28 | 4903 | |||
XM_011545777.2 | XP_011544079.1 | 14, 15 | 3166 | |||
XM_017023085.1 | XP_016878574.1 | 26, 27 | 4786 | |||
XM_017023086.1 | XP_016878575.1 | 26, 27 | 4786 | |||
XM_017023087.1 | XP_016878576.1 | 26, 27 | 4720 | |||
XM_017023088.1 | XP_016878577.1 | 23, 24 | 4507 | |||
GenBank mRNA | AB011128.2 | - | 27, 28 | 4831 | ||
AK295261.1 | - | 3, 4 | 422 | |||
AK315974.1 | - | 3, 4 | 422 | |||
BC150276.1 | - | 27, 28 | 4831 | 9007250 |

More Information
Other Species Targeted

