siRNA Details
Gene Symbol: OTUD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102653.1 | NP_001096123.1 | 21 | 3470 | ||
XM_005263079.4 | XP_005263136.1 | 21 | 3573 | |||
XM_011532040.2 | XP_011530342.2 | 21 | 3574 | |||
XM_011532041.2 | XP_011530343.2 | 21 | 3571 | |||
XM_011532042.2 | XP_011530344.1 | 21 | 3495 | |||
GenBank mRNA | AB028969.1 | - | 1 | 1965 | ||
AK302581.1 | - | 21 | 3432 | |||
BC057242.1 | - | 3 | 1718 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
