siRNA Details
Gene Symbol: 5730559C18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028872.3 | NP_083148.3 | 10 | 2728 | ||
GenBank mRNA | BC052416.1 | - | 10 | 2712 | 6402346 | |
BC053100.1 | - | 10 | 2712 | 6402346 | ||
BY388978.1 | - | 1 | 181 |
