siRNA Details
Gene Symbol: Casc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029617.2 | NP_083893.2 | 8 | 1153 | ||
GenBank mRNA | AK009772.1 | - | 8 | 1155 | ||
AK033132.1 | - | 8 | 1457 | |||
BC080815.1 | - | 8 | 1471 | 30104270 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
