siRNA Details
Gene Symbol: PCNX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014801.3 | NP_055616.3 | 25, 26 | 4579 | ||
XM_011544278.2 | XP_011542580.2 | 18, 19 | 2190 | |||
XM_017002391.1 | XP_016857880.1 | 24, 25 | 4143 | |||
XM_017002392.1 | XP_016857881.1 | 12, 13 | 1429 | |||
GenBank mRNA | AB007895.1 | - | 2, 3 | 1495 | ||
AK055374.1 | - | 6, 7 | 745 | |||
AK074078.1 | - | 20, 21 | 3651 | |||
AK090784.1 | - | 1, 2 | 811 | |||
AK307812.1 | - | 12, 13 | 1425 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
