siRNA Details
Gene Symbol: Yes1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033298.1 | NP_150640.1 | 3 | 535 | ||
XM_006245675.2 | XP_006245737.1 | 3 | 548 | |||
GenBank mRNA | AB037472.1 | - | 2 | 312 | ||
BC079403.1 | - | 3 | 535 | 7112783 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009535, NM_001205132, NM_001205133, X67677, AK167420, and others.
Gene Ontology Categories:
