siRNA Details
Gene Symbol: Adamts13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290464.1 | NP_001277393.1 | 24 | 3286 | ||
NM_001290465.1 | NP_001277394.1 | 23 | 3259 | |||
GenBank mRNA | AB071302.2 | - | 24 | 3286 | ||
EU034706.1 | - | 23 | 3138 |
Gene Symbol: BC094435 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK088963.1 | - | 1 | 3577 |
Gene Symbol: Cpne8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033851.1 | NP_001029023.1 | 4 | 496 | ||
GenBank mRNA | AK005311.1 | - | 4 | 430 | ||
BC048551.1 | - | 4 | 496 | |||
BC076564.1 | - | 4 | 455 |
Gene Symbol: Plekhh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC006045.1 | - | 2232 | |||
BC028900.1 | - | 2982 |
Gene Symbol: Sgip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK053400.1 | - | 2035 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
