siRNA Details
Gene Symbol: 4921509O09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014857.1 | - | 2 | 339 |
Gene Symbol: Gm20736 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037748.1 | NP_001032837.2 | 2 | 162 | ||
GenBank mRNA | BC100464.1 | - | 2 | 162 | 6705131 |
Gene Symbol: Gm20857 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BU945722.1 | - | 165 |
Gene Symbol: Sly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201530.2 | NP_963288.2 | 2 | 147 | ||
GenBank mRNA | BC049626.1 | - | 2 | 147 | 6705850 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
