siRNA Details
Gene Symbol: RABGAP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243765.1 | NP_001230694.1 | 2 | 636 | ||
NM_014857.4 | NP_055672.3 | 19 | 2520 | |||
XM_005245680.1 | XP_005245737.1 | 19 | 2450 | |||
XM_005245681.2 | XP_005245738.1 | 19 | 2409 | |||
XM_006711693.1 | XP_006711756.1 | 19 | 2450 | |||
XM_011510223.2 | XP_011508525.1 | 20 | 2477 | |||
XM_017002994.1 | XP_016858483.1 | 19 | 2407 | |||
XM_017002995.1 | XP_016858484.1 | 2 | 1508 | |||
XM_017002999.1 | XP_016858488.1 | 2 | 97 | |||
GenBank mRNA | AB007940.1 | - | 3 | 612 | ||
AB019490.1 | - | 4 | 353 | |||
AB019491.1 | - | 4 | 319 | |||
AB019492.2 | - | 3 | 208 | |||
AB019493.1 | - | 4 | 550 | |||
AB449904.1 | - | 18 | 2243 | |||
AK095838.2 | - | 19 | 2417 | |||
AY364260.1 | - | 12 | 1202 | |||
BC018630.2 | - | 3 | 219 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
