siRNA Details
Gene Symbol: Olr607 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000334.1 | NP_001000334.1 | 1 | 625 |
Gene Symbol: LOC100365887 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762003.1 | XP_008760225.1 | 2 | 505 |
Gene Symbol: LOC100912605 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749515.2 | XP_003749563.1 | 1 | 625 |
