siRNA Details
Gene Symbol: Olr841 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000405.1 | NP_001000405.1 | 1 | 121 |
Gene Symbol: LOC500460 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749941.1 | XP_003749989.1 | 1 | 121 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
