siRNA Details
Gene Symbol: PCNX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014801.3 | NP_055616.3 | 30, 31 | 5562 | ||
XM_011544278.2 | XP_011542580.2 | 23, 24 | 3173 | |||
XM_017002391.1 | XP_016857880.1 | 29, 30 | 5126 | |||
XM_017002392.1 | XP_016857881.1 | 17, 18 | 2412 | |||
GenBank mRNA | AB007895.1 | - | 7, 8 | 2478 | ||
AK055374.1 | - | 11, 12 | 1728 | |||
AK074078.1 | - | 25, 26 | 4634 | |||
AK090784.1 | - | 6, 7 | 1794 | |||
AK097229.1 | - | 16, 17 | 2137 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
