siRNA Details
Gene Symbol: Gtf2i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001512.2 | NP_001001512.2 | 24 | 2432 | ||
XM_006249162.2 | XP_006249224.1 | 28 | 2697 | |||
XM_006249163.2 | XP_006249225.1 | 28 | 2694 | |||
XM_006249164.2 | XP_006249226.1 | 27 | 2637 | |||
XM_006249165.2 | XP_006249227.1 | 27 | 2634 | |||
XM_006249166.2 | XP_006249228.1 | 27 | 2631 | |||
XM_006249167.2 | XP_006249229.1 | 26 | 2580 | |||
XM_006249168.2 | XP_006249230.1 | 26 | 2574 | |||
XM_006249169.2 | XP_006249231.1 | 26 | 2559 | |||
XM_006249170.2 | XP_006249232.1 | 28 | 2892 | |||
XM_006249171.2 | XP_006249233.1 | 25 | 2517 | |||
XM_006249172.2 | XP_006249234.1 | 25 | 2502 | |||
XM_006249173.2 | XP_006249235.1 | 25 | 2496 | |||
XM_006249174.2 | XP_006249236.1 | 24 | 2439 | |||
XM_008769141.1 | XP_008767363.1 | 28 | 2559 | |||
GenBank mRNA | AF547390.1 | - | 26 | 2469 | ||
AF547391.1 | - | 26 | 2469 | |||
BC085815.1 | - | 24 | 2570 | 7123295 |
