siRNA Details
Gene Symbol: LRRC37A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006607.2 | NP_001006608.2 | 1 | 1439 | ||
XM_011524841.2 | XP_011523143.1 | 1 | 1492 | |||
XM_011524842.2 | XP_011523144.1 | 1 | 1492 | |||
XM_011524843.2 | XP_011523145.1 | 1 | 1492 | |||
XM_011524844.2 | XP_011523146.1 | 1 | 1492 | |||
XM_011524846.2 | XP_011523148.1 | 1 | 1492 | |||
XM_011524848.2 | XP_011523150.1 | 1 | 1492 | |||
XM_011524849.1 | XP_011523151.1 | 1 | 1492 | |||
XM_011524850.2 | XP_011523152.1 | 1 | 1492 |
Gene Symbol: LRRC37A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014834.4 | NP_055649.4 | 1 | 1439 | ||
XM_005257879.4 | XP_005257936.1 | 1 | 1697 | |||
XM_005257880.4 | XP_005257937.1 | 1 | 1697 | |||
XM_005257882.4 | XP_005257939.1 | 2 | 1883 | |||
XM_005257883.4 | XP_005257940.1 | 1 | 1697 | |||
XM_005257884.4 | XP_005257941.1 | 1 | 1697 | |||
XM_006722210.3 | XP_006722273.1 | 1 | 1697 | |||
XM_006722211.3 | XP_006722274.1 | 1 | 1697 | |||
XM_011525539.2 | XP_011523841.1 | 1 | 1697 | |||
XM_017025472.1 | XP_016880961.1 | 3 | 1296 | |||
GenBank mRNA | BC040501.1 | - | 1 | 1439 | 5295808 | |
CR749232.1 | - | 1 | 1440 |
Gene Symbol: LRRC37A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199340.3 | NP_955372.2 | 4 | 2102 | ||
XM_017024593.1 | XP_016880082.1 | 2 | 1931 | |||
XM_017024594.1 | XP_016880083.1 | 2 | 1931 | |||
XM_017024595.1 | XP_016880084.1 | 2 | 1931 | |||
XM_017024596.1 | XP_016880085.1 | 2 | 1931 | |||
XM_017024597.1 | XP_016880086.1 | 2 | 1931 | |||
XM_017024598.1 | XP_016880087.1 | 2 | 1931 | |||
XM_017024599.1 | XP_016880088.1 | 3 | 1303 | |||
XM_017024600.1 | XP_016880089.1 | 2 | 1931 | |||
GenBank mRNA | AB011135.2 | - | 3 | 1967 | ||
BC043145.1 | - | 1 | 1444 | 5296147 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
