siRNA Details
Gene Symbol: ZNF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006956.2 | NP_008887.2 | 6 | 3107 | ||
NM_016265.3 | NP_057349.2 | 5 | 3221 | |||
GenBank mRNA | BC154414.1 | - | 5 | 3210 | 40148925 | |
BX640749.1 | - | 5 | 2630 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
