siRNA Details
Gene Symbol: AKAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004065.4 | NP_001004065.2 | 4 | 3449 | ||
NM_001136562.2 | NP_001130034.1 | 4 | 3287 | |||
NM_001198656.1 | NP_001185585.1 | 5 | 3488 | |||
GenBank mRNA | AK000306.1 | - | 2 | 2063 | ||
AK022693.1 | - | 3 | 1184 | |||
AL110268.1 | - | 3 | 1467 | |||
BC146863.1 | - | 4 | 3449 | 9056791 | ||
BC171800.1 | - | 5 | 3488 | 9054454 |
Gene Symbol: PALM2-AKAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007203.4 | NP_009134.1 | 11 | 4004 | ||
NM_147150.2 | NP_671492.1 | 10 | 3965 | |||
GenBank mRNA | AB023137.1 | - | 11 | 4004 | ||
AJ303079.1 | - | 11 | 3992 | |||
BC140818.1 | - | 10 | 3806 | 9021686 | ||
BX649126.1 | - | 9 | 3795 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
