siRNA Details
Gene Symbol: LILRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081637.2 | NP_001075106.2 | 15 | 2571 | ||
NM_001081638.3 | NP_001075107.2 | 15 | 2674 | |||
NM_001081639.3 | NP_001075108.2 | 15 | 2674 | |||
NM_001278398.2 | NP_001265327.2 | 14 | 2517 | |||
NM_006669.6 | NP_006660.4 | 16 | 2985 | |||
NR_103518.2 | 14 | 2489 | ||||
XM_011526331.2 | XP_011524633.1 | 14 | 2509 | |||
XM_011526332.2 | XP_011524634.1 | 14 | 2506 | |||
XM_011526335.2 | XP_011524637.1 | 13 | 2350 | |||
XM_011526336.2 | XP_011524638.1 | 13 | 2317 | |||
XM_017026182.1 | XP_016881671.1 | 14 | 2506 | |||
XM_017026183.1 | XP_016881672.1 | 14 | 2503 | |||
XM_017026184.1 | XP_016881673.1 | 14 | 2503 | |||
XM_017026185.1 | XP_016881674.1 | 14 | 2472 | |||
XM_017026186.1 | XP_016881675.1 | 15 | 2631 | |||
XM_017026187.1 | XP_016881676.1 | 16 | 2982 | |||
XM_017026188.1 | XP_016881677.1 | 15 | 2628 | |||
XM_017026189.1 | XP_016881678.1 | 15 | 2628 | |||
XM_017026190.1 | XP_016881679.1 | 16 | 2980 | |||
XM_017026191.1 | XP_016881680.1 | 13 | 2320 | |||
GenBank mRNA | AF283984.1 | - | 15 | 2578 | ||
AK094834.1 | - | 8 | 2184 | |||
BC015731.1 | - | 16 | 2740 | 4878915 |
Gene Symbol: AKR1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190906.1 | NP_001177835.1 | 8 | 2100 | ||
NM_001190907.1 | NP_001177836.1 | 8 | 2140 | |||
NM_005989.3 | NP_005980.1 | 9 | 2223 | |||
GenBank mRNA | Z28339.1 | - | 9 | 2205 |
Gene Symbol: ATP13A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011513124.2 | XP_011511426.1 | 34 | 5154 |
Gene Symbol: BIRC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DQ508248.1 | - | 3 | 235 | ||
DQ508249.1 | - | 3 | 255 |
Gene Symbol: CYB561D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX641103.1 | - | 1 | 1422 |
Gene Symbol: DOC2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282062.1 | NP_001268991.1 | 1 | 111 | ||
XM_011545975.1 | XP_011544277.1 | 3 | 584 | |||
XM_017023776.1 | XP_016879265.1 | 3 | 584 | |||
GenBank mRNA | BC063436.1 | - | 1 | 37 | 5394616 | |
DC399462.1 | - | 2 | 224 |
Gene Symbol: DPY19L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181787.2 | NP_861452.2 | 19 | 5040 | ||
XM_005250894.4 | XP_005250951.2 | 19 | 5253 | |||
GenBank mRNA | BC050303.1 | - | 1 | 2259 |
Gene Symbol: ERVFH21-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK023847.1 | - | 1 | 4916 |
Gene Symbol: GMPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011513263.2 | XP_011511565.1 | 16 | 5625 | ||
XM_017007422.1 | XP_016862911.1 | 18 | 6112 |
Gene Symbol: HES2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC012091.1 | - | 4 | 999 |
Gene Symbol: IDO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002164.5 | NP_002155.1 | 10 | 1672 | ||
GenBank mRNA | BF094375.1 | - | 1 | 195 |
Gene Symbol: KLHL22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK024369.1 | - | 1 | 1763 |
Gene Symbol: LOC100129373 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127907.1 | - | 1 | 2221 |
Gene Symbol: LRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052972.2 | NP_443204.1 | 2 | 1208 | ||
GenBank mRNA | AB073607.1 | - | 2 | 1220 | ||
AF403428.1 | - | 2 | 1208 | |||
AK027382.1 | - | 2 | 1220 | |||
AK303861.1 | - | 3 | 1169 | |||
BC034389.1 | - | 2 | 1209 | 5187278 | ||
BC070198.1 | - | 2 | 1265 | 4692618 |
Gene Symbol: NRG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017021937.1 | XP_016877426.1 | 12 | 1499 | ||
XM_017021938.1 | XP_016877427.1 | 11 | 3706 | |||
XM_017021939.1 | XP_016877428.1 | 11 | 1268 | |||
XM_017021940.1 | XP_016877429.1 | 10 | 4044 | |||
XM_017021941.1 | XP_016877430.1 | 8 | 1007 | |||
XM_017021942.1 | XP_016877431.1 | 8 | 1025 | |||
XM_017021943.1 | XP_016877432.1 | 8 | 1098 | |||
XM_017021944.1 | XP_016877433.1 | 9 | 3920 | |||
XM_017021946.1 | XP_016877435.1 | 10 | 3995 |
Gene Symbol: PLEKHH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB208939.1 | - | 11 | 3582 |
Gene Symbol: SAMMSON | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110000.1 | 4 | 1804 |
Gene Symbol: SMYD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017002094.1 | XP_016857583.1 | 1 | 2255 |
Gene Symbol: SP140L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC004921.2 | - | 6 | 1506 |
Gene Symbol: SRM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017002179.1 | XP_016857668.1 | 6 | 1021 |
Gene Symbol: TMEM50A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX537683.1 | - | 1 | 3012 |
Gene Symbol: TRIM64EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY010113.1 | - | 5 | 2406 |
Gene Symbol: UBIAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013319.2 | NP_037451.1 | 2 | 2947 | ||
GenBank mRNA | AK027433.1 | - | 1 | 1538 | ||
AK074487.1 | - | 1285 | ||||
AK074890.1 | - | 2 | 2940 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
