siRNA Details
Gene Symbol: PCLO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014510.2 | NP_055325.2 | 6 | 11212 | ||
NM_033026.5 | NP_149015.2 | 6 | 11212 | |||
GenBank mRNA | AK125999.1 | - | 1 | 569 | ||
BX649041.1 | - | 1 | 1084 | |||
CR749552.1 | - | 1 | 493 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
