siRNA Details
Gene Symbol: UBAP2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127320.2 | NP_001120792.1 | 25 | 3357 | ||
NM_001287815.1 | NP_001274744.1 | 25 | 3390 | |||
XM_005245673.3 | XP_005245730.1 | 25 | 3444 | |||
XM_005245674.1 | XP_005245731.1 | 25 | 3336 | |||
XM_017002989.1 | XP_016858478.1 | 25 | 3367 | |||
XM_017002990.1 | XP_016858479.1 | 25 | 3328 | |||
GenBank mRNA | AK124294.1 | - | 16 | 1667 | ||
D63478.1 | - | 25 | 3350 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
