siRNA Details
Gene Symbol: Wdr46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212491.2 | NP_997656.1 | 4 | 430 | ||
GenBank mRNA | BC085934.1 | - | 4 | 430 | 7112671 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wdr46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212491.2 | NP_997656.1 | 4 | 430 | ||
GenBank mRNA | BC085934.1 | - | 4 | 430 | 7112671 |