siRNA Details
Gene Symbol: DENND4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014856.2 | NP_055671.2 | 28 | 4949 | ||
XM_005245678.4 | XP_005245735.1 | 28 | 5106 | |||
XM_005245679.2 | XP_005245736.1 | 28 | 4956 | |||
XM_011510219.1 | XP_011508521.1 | 26 | 4355 | |||
XM_011510220.2 | XP_011508522.1 | 27 | 4769 | |||
XM_011510221.2 | XP_011508523.1 | 26 | 4477 | |||
XM_017002993.1 | XP_016858482.1 | 25 | 4331 | |||
GenBank mRNA | AB007945.2 | - | 28 | 4949 | ||
AK124049.1 | - | 3 | 1791 | |||
AK226053.1 | - | 23 | 3604 | |||
AL512734.1 | - | 1 | 132 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
