siRNA Details
Gene Symbol: RABGAP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243764.1 | NP_001230693.1 | 7 | 1385 | ||
NM_001243765.1 | NP_001230694.1 | 9 | 1993 | |||
XM_005245680.1 | XP_005245737.1 | 26 | 3807 | |||
XM_005245681.2 | XP_005245738.1 | 26 | 3766 | |||
XM_006711693.1 | XP_006711756.1 | 25 | 3609 | |||
XM_011510223.2 | XP_011508525.1 | 27 | 3834 | |||
XM_017002995.1 | XP_016858484.1 | 10 | 3079 | |||
XM_017002996.1 | XP_016858485.1 | 9 | 1562 | |||
XM_017002997.1 | XP_016858486.1 | 9 | 1670 | |||
XM_017002998.1 | XP_016858487.1 | 10 | 8657 | |||
XM_017002999.1 | XP_016858488.1 | 9 | 1470 | |||
GenBank mRNA | AB007940.1 | - | 10 | 1970 | ||
AB019492.2 | - | 10 | 1565 | |||
AK316355.1 | - | 7 | 1385 | |||
CR627374.1 | - | 8 | 1426 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
