siRNA Details
Gene Symbol: ZBTB40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083621.1 | NP_001077090.1 | 19 | 6316 | ||
NM_014870.3 | NP_055685.3 | 18 | 6028 | |||
XM_011542499.2 | XP_011540801.1 | 18 | 6051 | |||
XM_011542500.2 | XP_011540802.1 | 11 | 5301 | |||
XM_017003003.1 | XP_016858492.1 | 16 | 5692 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
