siRNA Details
Gene Symbol: Olr1568 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000532.1 | NP_001000532.1 | 63 |
Gene Symbol: LOC100911043 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751088.2 | XP_003751136.1 | 1 | 221 |
