siRNA Details
Gene Symbol: PUM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014878.4 | NP_055693.4 | 14 | 1424 | ||
GenBank mRNA | AF490254.1 | - | 11 | 908 | ||
AK225953.1 | - | 14 | 1389 | |||
AK292169.1 | - | 14 | 1367 | |||
AK315531.1 | - | 14 | 1403 | |||
AL832239.1 | - | 14 | 1390 | |||
AL832245.1 | - | 14 | 1381 | |||
BC016137.2 | - | 13 | 1328 | 3908516 | ||
CR456957.1 | - | 11 | 908 | |||
D13645.1 | - | 13 | 1326 |
