siRNA Details
Gene Symbol: Olr1523 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000713.1 | NP_001000713.1 | 136 | |||
XM_008768239.1 | XP_008766461.1 | 2 | 232 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr1523 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000713.1 | NP_001000713.1 | 136 | |||
XM_008768239.1 | XP_008766461.1 | 2 | 232 |