siRNA Details
Gene Symbol: Olr1622 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000838.1 | NP_001000838.1 | 1 | 23 |
Gene Symbol: LOC103690352 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770626.1 | XP_008768848.1 | 1 | 54 |
